Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271875.1 | NP_001258804.1 | 2, 3 | 757 | ||
NM_001292025.1 | NP_001278954.1 | 2, 3 | 1062 | |||
XM_006721900.2 | XP_006721963.1 | 2, 3 | 1372 | |||
XM_011524812.2 | XP_011523114.1 | 1, 2 | 96 | |||
XM_017024640.1 | XP_016880129.1 | 2, 3 | 727 | |||
XM_017024641.1 | XP_016880130.1 | 2, 3 | 725 | |||
GenBank mRNA | AK000746.1 | - | 2, 3 | 1062 | ||
AK000977.1 | - | 1, 2 | 496 | |||
BC056882.1 | - | 2, 3 | 757 | 6150503 |