Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGT1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003365.2 | 3 | 402 | |||
GenBank mRNA | BC020814.1 | - | 3 | 402 |
Gene Symbol: SUGT1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003667.3 | 3 | 625 | |||
GenBank mRNA | BE264023.1 | - | 3 | 506 | ||
DA804368.1 | - | 3 | 310 |