Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291980.1 | NP_001278909.1 | 6 | 884 | ||
NM_002011.4 | NP_002002.3 | 6 | 884 | |||
NM_022963.3 | NP_075252.2 | 5 | 722 | |||
NM_213647.2 | NP_998812.1 | 6 | 928 | |||
XM_005265838.3 | XP_005265895.1 | 6 | 836 | |||
XM_011534464.2 | XP_011532766.1 | 6 | 998 | |||
XM_011534465.2 | XP_011532767.1 | 6 | 921 | |||
GenBank mRNA | AB209631.1 | - | 3 | 2652 | ||
AF202063.1 | - | 5 | 705 | |||
AF359246.1 | - | 2 | 205 | |||
AK225280.1 | - | 6 | 842 | |||
AK225909.1 | - | 6 | 842 | |||
AK301169.1 | - | 6 | 851 | |||
BC011847.2 | - | 6 | 914 | 4121396 | ||
EU826603.1 | - | 5 | 669 | |||
JN007471.1 | - | 5 | 669 | |||
JN007472.1 | - | 5 | 669 | |||
JN007473.1 | - | 5 | 669 | |||
JN007474.1 | - | 5 | 669 | |||
JN007475.1 | - | 5 | 669 | |||
JN007476.1 | - | 5 | 669 | |||
JN007477.1 | - | 5 | 669 | |||
JN007478.1 | - | 5 | 669 | |||
JN007479.1 | - | 5 | 669 | |||
JN007480.1 | - | 5 | 669 | |||
JN007481.1 | - | 5 | 669 | |||
JN007482.1 | - | 5 | 669 | |||
L03840.1 | - | 6 | 839 | |||
X57205.1 | - | 5 | 724 |