Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDXP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020315.4 | NP_064711.1 | 2 | 814 | ||
GenBank mRNA | AK057265.1 | - | 2 | 380 | ||
AY125047.1 | - | 2 | 761 | |||
BC000320.2 | - | 2 | 813 | 2821743 | ||
BC009756.2 | - | 2 | 403 | |||
BC018844.2 | - | 2 | 782 | |||
BC064922.1 | - | 2 | 814 | 6141538 |