Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164664.1 | NP_001158136.1 | 28, 29 | 4269 | ||
NM_001290226.1 | NP_001277155.1 | 26, 27 | 3721 | |||
NM_001290227.1 | NP_001277156.1 | 25, 26 | 3292 | |||
NM_001297651.1 | NP_001284580.1 | 25, 26 | 3753 | |||
NM_015183.2 | NP_055998.1 | 27, 28 | 3462 | |||
XM_006714606.3 | XP_006714669.1 | 26, 27 | 3715 | |||
XM_006714610.2 | XP_006714673.1 | 24, 25 | 3501 | |||
XM_011543382.2 | XP_011541684.1 | 30, 31 | 4361 | |||
XM_011543384.2 | XP_011541686.1 | 25, 26 | 5420 | |||
XM_011543385.2 | XP_011541687.1 | 25, 26 | 6540 | |||
XM_011543386.2 | XP_011541688.1 | 25, 26 | 3371 | |||
XM_017009447.1 | XP_016864936.1 | 31, 32 | 4442 | |||
XM_017009448.1 | XP_016864937.1 | 29, 30 | 4325 | |||
XM_017009449.1 | XP_016864938.1 | 26, 27 | 3760 | |||
XM_017009450.1 | XP_016864939.1 | 26, 27 | 3454 | |||
XM_017009451.1 | XP_016864940.1 | 25, 26 | 3253 | |||
XM_017009452.1 | XP_016864941.1 | 26, 27 | 6769 | |||
XM_017009453.1 | XP_016864942.1 | 25, 26 | 13584 | |||
GenBank mRNA | AB002301.1 | - | 17, 18 | 2505 | ||
AY830839.1 | - | 27, 28 | 3463 | |||
BX648796.1 | - | 2, 3 | 300 | |||
BX648980.1 | - | 7, 8 | 1071 |