Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004356.2 | NP_001004356.1 | 7 | 1547 | ||
NM_001004358.1 | NP_001004358.1 | 7 | 1430 | |||
NM_021923.3 | NP_068742.2 | 6 | 1420 | |||
XM_011513486.1 | XP_011511788.1 | 7 | 1551 | |||
GenBank mRNA | AF279689.1 | - | 7 | 1430 | ||
AF312678.1 | - | 1526 | ||||
AJ277437.1 | - | 6 | 1420 | |||
AK022597.1 | - | 1 | 332 | |||
AK225486.1 | - | 6 | 1391 | |||
AK314365.1 | - | 7 | 1548 | |||
AY358303.1 | - | 1547 | ||||
BC015228.1 | - | 3 | 831 | |||
BC036769.1 | - | 7 | 1559 | 5744684 |