Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001158.3 | NP_001149.2 | 1 | 537 | ||
NM_009590.2 | NP_033720.2 | 1 | 537 | |||
GenBank mRNA | AF081363.1 | - | 1 | 528 | ||
BC142641.1 | - | 1 | 529 | 40147885 | ||
D88213.1 | - | 1 | 528 |