Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318144.1 | NP_001305073.1 | 3 | 443 | ||
NM_004925.4 | NP_004916.1 | 3 | 443 | |||
GenBank mRNA | AB001325.1 | - | 3 | 400 | ||
AJ493597.1 | - | 3 | 363 | |||
AK095363.1 | - | 3 | 713 | |||
AK292208.1 | - | 4 | 403 | |||
AK303206.1 | - | 3 | 403 | |||
AK315760.1 | - | 3 | 403 | |||
BC013566.1 | - | 3 | 377 | 3877096 | ||
BT007199.1 | - | 3 | 340 | |||
CR541991.1 | - | 4 | 340 | |||
CR542025.1 | - | 4 | 340 | |||
DA976040.1 | - | 3 | 443 |