Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007100.3 | NP_009031.1 | 1 | 112 | ||
NR_033743.1 | 1 | 112 | ||||
GenBank mRNA | AB028624.1 | - | 1 | 64 | ||
AK000656.1 | - | 1 | 27 | |||
BC003679.1 | - | 1 | 48 | 4052832 | ||
BC105610.1 | - | 1 | 54 | 6739801 | ||
BF215596.1 | - | 1 | 113 | |||
BG940181.1 | - | 1 | 40 | |||
BT006922.1 | - | 1 | 21 | |||
D50371.1 | - | 1 | 84 | |||
DA119852.1 | - | 1 | 104 |
Gene Symbol: MYL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713886.2 | XP_006713949.1 | 1 | 25 |