Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001196.3 | NP_001187.1 | 3 | 349 | ||
NM_001244567.1 | NP_001231496.1 | 3 | 260 | |||
NM_197966.2 | NP_932070.1 | 3 | 667 | |||
NM_197967.2 | NP_932071.1 | 2 | 279 | |||
XM_017028906.1 | XP_016884395.1 | 3 | 325 | |||
GenBank mRNA | AF042083.1 | - | 3 | 316 | ||
AF087891.1 | - | 3 | 314 | |||
AF250233.1 | - | 3 | 608 | |||
AK094795.1 | - | 3 | 608 | |||
AK313729.1 | - | 3 | 304 | |||
AY005151.1 | - | 3 | 266 | |||
BC009197.2 | - | 3 | 311 | 4025880 | ||
BC022072.1 | - | 3 | 349 | |||
BC033634.1 | - | 3 | 318 | 5538243 | ||
BC036364.1 | - | 3 | 642 | 4819949 | ||
BM561391.1 | - | 3 | 360 | |||
BM842561.1 | - | 2 | 279 | |||
BQ068464.1 | - | 3 | 327 | |||
CR407603.1 | - | 1 | 176 | |||
CR456389.1 | - | 2 | 218 | |||
D89803.1 | - | 2 | 249 | |||
DA158889.1 | - | 2 | 279 | |||
EU678292.1 | - | 3 | 266 | |||
EU678294.1 | - | 3 | 260 |