Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001196.3 | NP_001187.1 | 3, 4 | 390 | ||
NM_001244567.1 | NP_001231496.1 | 3, 4 | 301 | |||
NM_197966.2 | NP_932070.1 | 3, 4 | 708 | |||
NM_197967.2 | NP_932071.1 | 2, 3 | 320 | |||
XM_017028906.1 | XP_016884395.1 | 3, 4 | 366 | |||
GenBank mRNA | AF042083.1 | - | 3, 4 | 357 | ||
AF087891.1 | - | 3, 4 | 355 | |||
AF250233.1 | - | 3, 4 | 649 | |||
AK094795.1 | - | 3, 4 | 649 | |||
AK313729.1 | - | 3, 4 | 345 | |||
BC009197.2 | - | 3, 4 | 352 | 4025880 | ||
BC022072.1 | - | 3, 4 | 390 | |||
BC033634.1 | - | 3, 4 | 359 | 5538243 | ||
BC036364.1 | - | 3, 4 | 683 | 4819949 | ||
BM561391.1 | - | 3, 4 | 401 | |||
BM842561.1 | - | 2, 3 | 320 | |||
BQ068464.1 | - | 3, 4 | 368 | |||
CR407603.1 | - | 1, 2 | 217 | |||
CR456389.1 | - | 2, 3 | 259 | |||
D89803.1 | - | 2, 3 | 290 | |||
DA158889.1 | - | 2, 3 | 320 | |||
EU447348.1 | - | 1, 2 | 40 | |||
EU678294.1 | - | 3, 4 | 301 |