Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032515.4 | NP_115904.1 | 4, 5 | 807 | ||
XM_011511696.2 | XP_011509998.1 | 4, 5 | 611 | |||
XM_017004775.1 | XP_016860264.1 | 5, 6 | 909 | |||
XM_017004776.1 | XP_016860265.1 | 5, 6 | 835 | |||
GenBank mRNA | AF174487.1 | - | 4, 5 | 656 | ||
BC006203.2 | - | 4, 5 | 631 | |||
BT007272.1 | - | 3, 4 | 515 | |||
BX372066.2 | - | 2, 3 | 466 | |||
BX460019.2 | - | 4, 5 | 809 | |||
CN279175.1 | - | 1, 2 | 100 |