Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021950.3 | NP_068769.2 | 6 | 747 | ||
NM_152866.2 | NP_690605.1 | 7 | 1010 | |||
GenBank mRNA | AK292168.1 | - | 6 | 883 | ||
BC002807.2 | - | 6 | 697 | 3634040 | ||
X07203.1 | - | 6 | 684 | |||
X12530.1 | - | 6 | 687 |