Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021950.3 | NP_068769.2 | 7 | 929 | ||
NM_152866.2 | NP_690605.1 | 8 | 1192 | |||
GenBank mRNA | AK225630.1 | - | 7 | 852 | ||
AK292168.1 | - | 7 | 1065 | |||
AK300025.1 | - | 4 | 386 | |||
BC002807.2 | - | 7 | 879 | 3634040 | ||
X07203.1 | - | 7 | 866 | |||
X12530.1 | - | 7 | 869 |