Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005507.2 | NP_005498.1 | 2 | 525 | ||
GenBank mRNA | AK097690.1 | - | 3 | 1007 | ||
BC011005.2 | - | 2 | 353 | 4079579 | ||
BC012265.1 | - | 2 | 358 | 3862540 | ||
BC012318.1 | - | 2 | 357 | 3538185 | ||
BC018256.2 | - | 2 | 353 | 3448134 | ||
BC024975.1 | - | 1 | 60 | |||
BT006846.1 | - | 1 | 291 | |||
D00682.1 | - | 1 | 291 | |||
EU794603.1 | - | 1 | 291 | |||
U21909.1 | - | 1 | 291 | |||
X95404.1 | - | 2 | 342 |