Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303499.1 | NP_001290428.1 | 7 | 1146 | ||
NM_001303501.1 | NP_001290430.1 | 7 | 1242 | |||
NM_004368.3 | NP_004359.1 | 7 | 1179 | |||
NM_201277.2 | NP_958434.1 | 6 | 1062 | |||
GenBank mRNA | AK024835.1 | - | 7 | 825 | ||
AK057960.1 | - | 6 | 708 | |||
AK093630.1 | - | 1 | 964 | |||
AK126391.1 | - | 6 | 727 | |||
AK222924.1 | - | 7 | 815 | |||
AK293171.1 | - | 7 | 792 | |||
AK295271.1 | - | 7 | 795 | |||
AK297774.1 | - | 8 | 764 | |||
AK300788.1 | - | 7 | 877 | |||
BC141818.1 | - | 7 | 822 | 40148129 | ||
BC141833.1 | - | 7 | 822 | 40148834 | ||
BC148264.1 | - | 7 | 814 | 8860204 | ||
BM976651.1 | - | 1 | 549 | |||
BQ876824.1 | - | 7 | 754 | |||
BU844400.1 | - | 3 | 401 | |||
CR541810.1 | - | 7 | 792 | |||
CR541831.1 | - | 7 | 792 | |||
D83735.1 | - | 7 | 819 |