Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL9A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001852.3 | NP_001843.1 | 2, 3 | 214 | ||
XM_006710365.3 | XP_006710428.1 | 3, 4 | 322 | |||
XM_011540715.2 | XP_011539017.1 | 2, 3 | 790 | |||
XM_011540717.2 | XP_011539019.1 | 2, 3 | 787 | |||
XM_017000332.1 | XP_016855821.1 | 2, 3 | 256 | |||
GenBank mRNA | BC041479.1 | - | 2, 3 | 104 | ||
BC136326.1 | - | 2, 3 | 207 | 9020313 | ||
BC136327.1 | - | 2, 3 | 207 | 9020314 | ||
M95610.1 | - | 2, 3 | 103 |