Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000756.3 | NP_000747.1 | 2 | 940 | ||
XM_017013090.1 | XP_016868579.1 | 6 | 1278 | |||
XM_017013091.1 | XP_016868580.1 | 6 | 1341 | |||
XM_017013092.1 | XP_016868581.1 | 6 | 1279 | |||
XM_017013093.1 | XP_016868582.1 | 6 | 1418 | |||
GenBank mRNA | AK075431.1 | - | 2 | 886 | ||
BC002599.2 | - | 2 | 751 | 3160933 | ||
BC011031.1 | - | 2 | 761 | 4280754 | ||
BT007453.1 | - | 1 | 573 | |||
KU177994.1 | - | 1 | 573 | |||
KU177995.1 | - | 2 | 400 |