Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CST6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323.3 | NP_001314.1 | 2 | 312 | ||
GenBank mRNA | AA778147.1 | - | 2 | 304 | ||
AI127639.1 | - | 2 | 312 | |||
AK092391.1 | - | 1 | 1354 | |||
BC031334.1 | - | 2 | 283 | 5446833 | ||
CR456872.1 | - | 2 | 258 | |||
U62800.1 | - | 2 | 281 | |||
U81233.1 | - | 2 | 276 |