Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTBP2 SIRNA Assays ›
Gene Symbol:
CTBP2
Gene Name:
C-terminal binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 124987849 - 125161061 on Build GRCh38
UniGene ID:
Hs.501345
Species:
Human
siRNA ID
HSS141738
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083914.2
NP_001077383.1
6, 7
993
NM_001290214.2
NP_001277143.1
6, 7
978
NM_001290215.2
NP_001277144.1
6, 7
1242
NM_001321012.1
NP_001307941.1
6, 7
937
NM_001321013.1
NP_001307942.1
5, 6
1138
NM_001321014.1
NP_001307943.1
6, 7
1239
NM_001329.3
NP_001320.1
6, 7
981
NM_022802.2
NP_073713.2
4, 5
2298
XM_005269561.2
XP_005269618.1
7, 8
1164
XM_005269564.2
XP_005269621.1
7, 8
1372
XM_005269567.2
XP_005269624.1
7, 8
1371
XM_005269568.4
XP_005269625.1
7, 8
1099
XM_005269569.2
XP_005269626.1
6, 7
1060
XM_005269571.2
XP_005269628.1
5, 6
819
XM_005269572.3
XP_005269629.1
5, 6
867
XM_006717642.2
XP_006717705.1
8, 9
1293
XM_011539349.2
XP_011537651.1
4, 5
1075
XM_011539351.1
XP_011537653.1
9, 10
1394
XM_011539353.1
XP_011537655.1
8, 9
1265
XM_011539354.1
XP_011537656.1
7, 8
1189
XM_011539355.1
XP_011537657.1
6, 7
1267
XM_011539357.1
XP_011537659.1
6, 7
1005
XM_011539358.1
XP_011537660.1
5, 6
1083
XM_017015756.1
XP_016871245.1
7, 8
1035
XM_017015757.1
XP_016871246.1
6, 7
2761
GenBank mRNA
AF016507.1
-
6, 7
898
AF222711.1
-
4, 5
2315
AK290390.1
-
6, 7
842
AL833398.1
-
6, 7
953
BC002486.2
-
6, 7
954
BC037900.2
-
4, 5
962
5276086
BC047018.1
-
6, 7
958
BC052276.1
-
6, 7
935
BC072020.1
-
6, 7
915
BM800106.1
-
4, 5
653
BT007012.1
-
4, 5
553
CV029885.1
-
4, 5
553
DB200827.1
-
4, 5
526
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of cell proliferation
viral genome replication
positive regulation of chromatin binding
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of retinoic acid receptor signaling pathway
white fat cell differentiation
oxidation-reduction process
chromatin binding
transcription coactivator activity
transcription corepressor activity
protein binding
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
protein homodimerization activity
retinoic acid receptor binding
NAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0