Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001933.4 | NP_001924.2 | 12 | 983 | ||
NR_033814.1 | 11 | 925 | ||||
NR_045209.1 | 11 | 934 | ||||
GenBank mRNA | AK289414.1 | - | 12 | 981 | ||
AK299505.1 | - | 9 | 817 | |||
AK316463.1 | - | 11 | 900 | |||
AL538322.3 | - | 5 | 423 | |||
BC000302.2 | - | 12 | 945 | 2820348 | ||
BC001922.1 | - | 12 | 945 | 2820348 | ||
BX248276.1 | - | 11 | 902 | |||
BX248764.1 | - | 10 | 814 | |||
CR456727.1 | - | 12 | 920 | |||
D16373.1 | - | 12 | 932 | |||
L37418.1 | - | 12 | 920 |