Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMD SIRNA Assays ›
Gene Symbol:
DMD
Gene Name:
dystrophin
Gene Aliases:
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
Chromosome Location:
Chr.X: 31119219 - 33339609 on Build GRCh38
UniGene ID:
Hs.495912
Species:
Human
siRNA ID
HSS141852
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000109.3
NP_000100.2
64
9650
NM_004006.2
NP_003997.1
64
9574
NM_004009.3
NP_004000.1
64
9581
NM_004010.3
NP_004001.1
64
9664
NM_004011.3
NP_004002.2
36
5352
NM_004012.3
NP_004003.1
36
5495
NM_004013.2
NP_004004.1
21
2991
NM_004014.2
NP_004005.1
10
1204
NM_004015.2
NP_004006.1
3
204
NM_004016.2
NP_004007.1
3
204
NM_004017.2
NP_004008.1
3
204
NM_004018.2
NP_004009.1
3
204
NM_004019.2
NP_004010.1
3
204
NM_004020.3
NP_004011.2
21
2991
NM_004021.2
NP_004012.1
21
2991
NM_004022.2
NP_004013.1
21
2991
NM_004023.2
NP_004014.1
21
2991
XM_006724468.2
XP_006724531.1
64
9527
XM_006724469.3
XP_006724532.1
64
9501
XM_006724470.3
XP_006724533.1
64
9527
XM_006724473.2
XP_006724536.1
63
9389
XM_006724474.3
XP_006724537.1
64
9527
XM_006724475.2
XP_006724538.1
64
9527
XM_011545467.1
XP_011543769.1
63
9404
XM_011545468.2
XP_011543770.1
64
9527
XM_017029328.1
XP_016884817.1
64
9527
XM_017029331.1
XP_016884820.1
24
4103
GenBank mRNA
AB208836.1
-
18
2438
AK299936.1
-
3
246
BC028720.1
-
3
255
BC070078.1
-
3
205
BC094758.1
-
3
218
BC127103.1
-
18
2515
BC150141.1
-
18
2515
DB233002.1
-
3
204
KU178030.1
-
3
126
KU178031.1
-
3
126
KU178032.1
-
3
126
KU178033.1
-
3
126
KU178034.1
-
3
126
M18533.1
-
64
9538
M92650.1
-
3
178
S38776.1
-
3
249
X14298.1
-
64
9428
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
cytoskeleton organization
muscle organ development
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
regulation of skeletal muscle contraction
muscle attachment
muscle filament sliding
negative regulation of peptidyl-serine phosphorylation
cellular protein localization
response to muscle stretch
peptide biosynthetic process
cellular protein complex assembly
muscle cell cellular homeostasis
muscle fiber development
cardiac muscle contraction
regulation of ryanodine-sensitive calcium-release channel activity
cardiac muscle cell action potential
regulation of voltage-gated calcium channel activity
negative regulation of peptidyl-cysteine S-nitrosylation
positive regulation of sodium ion transmembrane transporter activity
dystroglycan binding
actin binding
structural constituent of cytoskeleton
protein binding
zinc ion binding
structural constituent of muscle
myosin binding
vinculin binding
nitric-oxide synthase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0