Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004952.4 | NP_004943.1 | 3 | 547 | ||
GenBank mRNA | AK296030.1 | - | 3 | 531 | ||
AK316248.1 | - | 3 | 510 | |||
BC017722.1 | - | 3 | 547 | 4397263 | ||
BC110406.1 | - | 3 | 537 | 5732044 | ||
L37360.1 | - | 3 | 483 | |||
U14187.1 | - | 3 | 534 |