Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258012.3 | NP_001244941.2 | 5 | 702 | ||
NM_001258013.3 | NP_001244942.2 | 6 | 666 | |||
NM_001258014.3 | NP_001244943.2 | 5 | 549 | |||
NM_001258015.2 | NP_001244944.1 | 4 | 453 | |||
NM_001258016.3 | NP_001244945.2 | 4 | 518 | |||
NM_004110.5 | NP_004101.3 | 5 | 573 | |||
NM_024417.4 | NP_077728.3 | 5 | 573 | |||
NR_047576.2 | 5 | 673 | ||||
XM_006721772.1 | XP_006721835.1 | 5 | 521 | |||
XM_011524527.1 | XP_011522829.1 | 5 | 521 | |||
XM_011524528.1 | XP_011522830.1 | 5 | 610 | |||
XM_011524529.1 | XP_011522831.1 | 5 | 613 | |||
XM_011524531.2 | XP_011522833.1 | 4 | 506 | |||
XM_011524532.2 | XP_011522834.1 | 4 | 513 | |||
XM_011524533.2 | XP_011522835.1 | 4 | 499 | |||
GenBank mRNA | AK094120.1 | - | 6 | 771 | ||
AK098648.1 | - | 5 | 510 | |||
AK223530.1 | - | 5 | 503 | |||
AK293208.1 | - | 6 | 611 | |||
AK293210.1 | - | 5 | 610 | |||
AK295307.1 | - | 5 | 702 | |||
AK298908.1 | - | 4 | 396 | |||
AK301779.1 | - | 5 | 486 | |||
AK301977.1 | - | 4 | 518 | |||
BC002960.1 | - | 5 | 512 | 3537008 | ||
BC018911.2 | - | 5 | 519 | |||
BC034763.1 | - | 5 | 1174 | |||
BC063493.1 | - | 5 | 511 | 5404814 | ||
BT006684.1 | - | 5 | 486 | |||
J03826.1 | - | 5 | 506 |