Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002032.2 | NP_002023.2 | 4 | 904 | ||
GenBank mRNA | AB062402.1 | - | 4 | 857 | ||
AY258285.1 | - | 1462 | ||||
BC000857.1 | - | 4 | 863 | 3459353 | ||
BC001399.2 | - | 4 | 832 | 3051458 | ||
BC011359.2 | - | 4 | 854 | 4157296 | ||
BC013724.2 | - | 4 | 857 | 3857790 | ||
BC015156.1 | - | 4 | 727 | 3883694 | ||
BC016009.1 | - | 4 | 879 | 4666545 | ||
BC016857.1 | - | 4 | 823 | 3848510 | ||
BC020300.1 | - | 4 | 840 | |||
BC035441.1 | - | 4 | 863 | |||
BC063514.1 | - | 4 | 846 | 3882345 | ||
BC066341.1 | - | 5 | 781 | 5288688 | ||
BC066961.1 | - | 4 | 877 | 4796312 | ||
BC070494.1 | - | 5 | 818 | |||
BC073750.1 | - | 4 | 861 | 6256153 | ||
BC104643.1 | - | 4 | 879 | 4043391 | ||
BC105802.1 | - | 5 | 856 | |||
BM971420.1 | - | 1 | 338 | |||
L20941.1 | - | 4 | 877 | |||
M11146.1 | - | 4 | 746 | |||
M12937.1 | - | 4 | 693 | |||
M97164.1 | - | 4 | 798 | |||
X00318.1 | - | 4 | 757 |
Gene Symbol: BEST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA388926.1 | - | 1 | 151 |