Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000157.3 | NP_000148.2 | 3 | 456 | ||
NM_001005741.2 | NP_001005741.1 | 4 | 715 | |||
NM_001005742.2 | NP_001005742.1 | 4 | 696 | |||
NM_001171811.1 | NP_001165282.1 | 2 | 532 | |||
NM_001171812.1 | NP_001165283.1 | 3 | 453 | |||
GenBank mRNA | AK291911.1 | - | 4 | 694 | ||
AK298900.1 | - | 3 | 453 | |||
AK300186.1 | - | 3 | 421 | |||
AK300829.1 | - | 2 | 369 | |||
AK300876.1 | - | 4 | 696 | |||
AK301879.1 | - | 3 | 453 | |||
AK302000.1 | - | 2 | 281 | |||
AK311242.1 | - | 4 | 304 | |||
AK312502.1 | - | 4 | 512 | |||
AL547558.3 | - | 5 | 627 | |||
AU122729.1 | - | 5 | 616 | |||
AU131071.1 | - | 4 | 524 | |||
AU140959.1 | - | 3 | 440 | |||
AU141794.1 | - | 4 | 531 | |||
BC003356.1 | - | 3 | 395 | 2899915 | ||
BX382279.2 | - | 5 | 614 | |||
BX648487.1 | - | 4 | 524 | |||
D13286.1 | - | 3 | 395 | |||
DC297079.1 | - | 3 | 456 | |||
K02920.1 | - | 2 | 275 | |||
KJ690771.1 | - | 3 | 339 | |||
KJ690773.1 | - | 3 | 339 | |||
M16328.1 | - | 3 | 725 | |||
M19285.1 | - | 3 | 412 |