Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000159.3 | NP_000150.1 | 5, 6 | 442 | ||
NM_013976.3 | NP_039663.1 | 5, 6 | 442 | |||
NR_102317.1 | 4, 5 | 750 | ||||
XM_006722721.2 | XP_006722784.1 | 5, 6 | 411 | |||
XM_011527899.2 | XP_011526201.1 | 5, 6 | 411 | |||
XM_011527900.2 | XP_011526202.1 | 5, 6 | 411 | |||
XM_017026580.1 | XP_016882069.1 | 5, 6 | 411 | |||
GenBank mRNA | AK098370.1 | - | 5, 6 | 411 | ||
AK290407.1 | - | 5, 6 | 411 | |||
AK300841.1 | - | 4, 5 | 699 | |||
AK309892.1 | - | 4, 5 | 699 | |||
AU135567.1 | - | 5, 6 | 411 | |||
BC002579.2 | - | 5, 6 | 395 | 3142978 | ||
BT006706.1 | - | 4, 5 | 334 | |||
U69141.1 | - | 4, 5 | 370 |