Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310220.1 | NP_001297149.1 | 3 | 248 | ||
NM_001501.1 | NP_001492.1 | 3 | 253 | |||
NM_178331.1 | NP_847901.1 | 3 | 232 | |||
NM_178332.1 | NP_847902.1 | 3 | 229 | |||
XM_017027823.1 | XP_016883312.1 | 3 | 2839 | |||
GenBank mRNA | AF036330.1 | - | 3 | 253 | ||
AF190640.1 | - | 2 | 103 | |||
BC069362.1 | - | 3 | 252 | 7262673 | ||
BC115399.1 | - | 3 | 231 | 40069240 | ||
BC115400.1 | - | 3 | 247 | 40069241 | ||
BF061974.1 | - | 2 | 167 | |||
BF062277.1 | - | 2 | 167 |