Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286099.1 | NP_001273028.1 | 3 | 1219 | ||
NM_005284.4 | NP_005275.1 | 2 | 1110 | |||
GenBank mRNA | AK295250.1 | - | 3 | 1164 | ||
BC043414.2 | - | 1 | 898 | 5294842 | ||
BC117298.1 | - | 1 | 996 | 40125849 | ||
BC126301.1 | - | 1 | 998 | 8992017 | ||
HQ709190.1 | - | 1 | 880 |
Set Membership: |
G-protein coupled receptor |