Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001512.3 | NP_001503.1 | 4, 5 | 412 | ||
XM_005249035.3 | XP_005249092.1 | 4, 5 | 505 | |||
XM_011514534.2 | XP_011512836.1 | 3, 4 | 355 | |||
XM_011514535.2 | XP_011512837.1 | 3, 4 | 451 | |||
GenBank mRNA | AF020918.1 | - | 4, 5 | 332 | ||
AF025887.1 | - | 4, 5 | 335 | |||
AF125271.1 | - | 3, 4 | 262 | |||
AF125272.1 | - | 3, 4 | 262 | |||
AF125273.1 | - | 3, 4 | 262 | |||
AK054804.1 | - | 2, 3 | 769 | |||
AK315369.1 | - | 4, 5 | 311 | |||
BC015523.2 | - | 3, 4 | 282 | 3915425 | ||
BG716171.1 | - | 4, 5 | 379 | |||
Y13047.1 | - | 3, 4 | 262 |