Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021784.4 | NP_068556.2 | 1 | 246 | ||
NM_153675.2 | NP_710141.1 | 2 | 233 | |||
GenBank mRNA | AB028021.1 | - | 2 | 239 | ||
AK301611.1 | - | 1 | 46 | |||
BC006545.2 | - | 1 | 39 | |||
BC011780.2 | - | 1 | 85 | 3940139 | ||
BC019288.1 | - | 1 | 39 | |||
BM273279.1 | - | 1 | 253 | |||
BQ643619.1 | - | 234 |