Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195517.1 | NP_001182446.1 | 1 | 745 | ||
NM_005521.3 | NP_005512.1 | 1 | 745 | |||
XM_011539744.2 | XP_011538046.1 | 1 | 1261 | |||
XM_011539745.2 | XP_011538047.1 | 1 | 1276 | |||
GenBank mRNA | BC130530.1 | - | 1 | 745 | 40146561 | |
M62626.1 | - | 1 | 1525 | |||
M75952.1 | - | 1 | 519 | |||
S38742.1 | - | 1 | 605 |