Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039132.2 | NP_001034221.1 | 3 | 754 | ||
NM_001544.4 | NP_001535.1 | 3 | 831 | |||
NM_022377.3 | NP_071772.1 | 2 | 978 | |||
GenBank mRNA | BC000046.2 | - | 3 | 747 | 3505509 | |
BC029364.1 | - | 3 | 831 | 4659161 | ||
BT009816.1 | - | 3 | 708 | |||
L27670.1 | - | 2 | 938 | |||
L27671.1 | - | 3 | 791 |