Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021019.4 | NP_066299.2 | 4 | 400 | ||
NM_079423.3 | NP_524147.2 | 4 | 400 | |||
GenBank mRNA | AB046613.1 | - | 4 | 299 | ||
AK301198.1 | - | 3 | 578 | |||
BC006781.2 | - | 2 | 230 | |||
BC017455.1 | - | 4 | 287 | 3538373 | ||
BC071661.1 | - | 4 | 283 | 5561575 | ||
BC093066.1 | - | 4 | 296 | 30722094 | ||
CA427580.1 | - | 4 | 424 | |||
CN401857.1 | - | 4 | 284 | |||
CR456839.1 | - | 2 | 154 | |||
CR457014.1 | - | 3 | 259 | |||
M22918.1 | - | 4 | 290 | |||
M22920.1 | - | 3 | 268 | |||
M31212.1 | - | 4 | 299 | |||
U02629.1 | - | 4 | 297 |