Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002493.4 | NP_002484.1 | 2 | 353 | ||
NM_182739.2 | NP_877416.1 | 2 | 353 | |||
GenBank mRNA | AF035840.1 | - | 2 | 297 | ||
AF067167.1 | - | 2 | 326 | |||
AK289702.1 | - | 2 | 307 | |||
BC009801.1 | - | 2 | 332 | 4334414 | ||
BC016708.1 | - | 2 | 367 | |||
BC029247.1 | - | 2 | 298 | 5171627 | ||
BI669356.1 | - | 2 | 332 | |||
BP208846.1 | - | 2 | 351 | |||
BU952486.1 | - | 3 | 506 | |||
CB140469.1 | - | 2 | 354 | |||
CR456920.1 | - | 2 | 229 |