Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184979.1 | NP_001171908.1 | 2 | 279 | ||
NM_004552.2 | NP_004543.1 | 2 | 283 | |||
GenBank mRNA | AF020352.1 | - | 2 | 233 | ||
AF047434.1 | - | 2 | 244 | |||
AF086268.1 | - | 2 | 234 | |||
AI244607.1 | - | 2 | 298 | |||
AK312188.1 | - | 2 | 257 | |||
BC001884.1 | - | 2 | 201 | 3544747 | ||
BC093057.1 | - | 2 | 223 | 30382835 | ||
CR456904.1 | - | 1 | 173 |