Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002515.2 | NP_002506.2 | 2 | 533 | ||
NM_006489.2 | NP_006480.2 | 2 | 533 | |||
NM_006491.2 | NP_006482.1 | 2 | 533 | |||
XM_017021342.1 | XP_016876831.1 | 2 | 589 | |||
XM_017021343.1 | XP_016876832.1 | 2 | 589 | |||
XM_017021344.1 | XP_016876833.1 | 2 | 298 | |||
XM_017021345.1 | XP_016876834.1 | 2 | 747 | |||
XM_017021346.1 | XP_016876835.1 | 2 | 747 | |||
XM_017021347.1 | XP_016876836.1 | 2 | 628 | |||
XM_017021349.1 | XP_016876838.1 | 2 | 748 | |||
GenBank mRNA | AK226114.1 | - | 2 | 250 | ||
AK289639.1 | - | 2 | 219 | |||
AK291022.1 | - | 2 | 356 | |||
AK291468.1 | - | 2 | 234 | |||
AK301526.1 | - | 2 | 215 | |||
BC075038.2 | - | 2 | 238 | 30915356 | ||
BC075039.2 | - | 2 | 239 | 30915572 | ||
CN409914.1 | - | 2 | 536 | |||
U04840.1 | - | 2 | 275 |