Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002608.3 | NP_002599.1 | 5 | 1573 | ||
NM_033016.3 | NP_148937.1 | 5 | 473 | |||
GenBank mRNA | BC029822.1 | - | 5 | 793 | 5174750 | |
BC077725.1 | - | 5 | 1305 | 6379783 | ||
CR456538.1 | - | 5 | 489 | |||
CR541807.1 | - | 5 | 481 | |||
M12783.1 | - | 5 | 1573 | |||
X02744.1 | - | 5 | 598 | |||
X02811.1 | - | 5 | 1463 | |||
X03702.1 | - | 4 | 406 | |||
X83705.1 | - | 5 | 473 |