Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301130.1 | NP_001288059.1 | 2, 3 | 217 | ||
NM_001301131.1 | NP_001288060.1 | 2, 3 | 217 | |||
NM_021974.4 | NP_068809.1 | 2, 3 | 217 | |||
XM_017028832.1 | XP_016884321.1 | 2, 3 | 183 | |||
GenBank mRNA | BC003582.1 | - | 2, 3 | 195 | 3546712 | |
BP195499.1 | - | 2, 3 | 183 | |||
CR456546.1 | - | 2, 3 | 107 | |||
CR457021.1 | - | 2, 3 | 92 | |||
DA372532.1 | - | 2, 3 | 217 | |||
DN989996.1 | - | 2, 3 | 192 |