Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POU5F1B SIRNA Assays ›
Gene Symbol:
POU5F1B
Gene Name:
POU class 5 homeobox 1B
Gene Aliases:
OCT4-PG1, OCT4PG1, OTF3C, OTF3P1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8
Chromosome Location:
Chr.8: 127415612 - 127417210 on Build GRCh38
UniGene ID:
Hs.450254
Species:
Human
siRNA ID
HSS143406
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POU5F1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159542.1
NP_001153014.1
1
1020
GenBank mRNA
AF268615.1
-
1
765
AF268618.1
-
1
765
DQ486513.1
-
1
1020
GU480835.1
-
1
766
GU480840.1
-
1
766
GU480841.1
-
1
766
GU480842.1
-
1
766
GU480845.1
-
1
766
GU480849.1
-
1
768
GU480851.1
-
1
767
GU480864.1
-
1
766
GU480868.1
-
1
766
GU480878.1
-
1
766
GU480880.1
-
1
766
GU480883.1
-
1
766
GU480884.1
-
1
765
GU480885.1
-
1
767
GU480886.1
-
1
766
GU480889.1
-
1
766
GU480896.1
-
1
766
GU480899.1
-
1
767
GU480902.1
-
1
766
GU480905.1
-
1
766
GU480906.1
-
1
766
GU480908.1
-
1
766
GU480910.1
-
1
768
GU480911.1
-
1
766
GU480912.1
-
1
766
GU480913.1
-
1
766
HQ122710.1
-
1
766
HQ122711.1
-
1
766
HQ122712.1
-
1
766
HQ122713.1
-
1
766
HQ122714.1
-
1
766
HQ122715.1
-
1
765
HQ122716.1
-
1
766
HQ122717.1
-
1
766
HQ122718.1
-
1
766
HQ122719.1
-
1
766
HQ122720.1
-
1
766
HQ122721.1
-
1
766
HQ122749.1
-
1
766
HQ122750.1
-
1
766
HQ122751.1
-
1
766
HQ122752.1
-
1
766
HQ122781.1
-
1
766
HQ122782.1
-
1
766
HQ122783.1
-
1
766
HQ122784.1
-
1
766
HQ122785.1
-
1
766
HQ122786.1
-
1
766
HQ122787.1
-
1
766
HQ122788.1
-
1
766
HQ122789.1
-
1
766
HQ122790.1
-
1
766
HQ122816.1
-
1
766
HQ122817.1
-
1
766
HQ122818.1
-
1
766
HQ122819.1
-
1
766
HQ122820.1
-
1
766
HQ122821.1
-
1
766
HQ122822.1
-
1
766
HQ122823.1
-
1
766
HQ122824.1
-
1
766
HQ122825.1
-
1
766
HQ122826.1
-
1
766
HQ122827.1
-
1
766
HQ122841.1
-
1
766
HQ122842.1
-
1
766
HQ122843.1
-
1
766
HQ122844.1
-
1
766
HQ122845.1
-
1
766
HQ122846.1
-
1
766
HQ122848.1
-
1
766
HQ122849.1
-
1
766
HQ122850.1
-
1
767
HQ122851.1
-
1
766
HQ122852.1
-
1
766
HQ122853.1
-
1
766
HQ122854.1
-
1
766
HQ122855.1
-
1
766
HQ122856.1
-
1
766
HQ122857.1
-
1
766
HQ122863.1
-
1
765
HQ122864.1
-
1
766
HQ122866.1
-
1
766
HQ122867.1
-
1
766
HQ122868.1
-
1
766
HQ122869.1
-
1
766
HQ122870.1
-
1
766
HQ122871.1
-
1
766
HQ122872.1
-
1
765
HQ122874.1
-
1
766
HQ122875.1
-
1
766
HQ122877.1
-
1
766
HQ122878.1
-
1
766
HQ122879.1
-
1
766
HQ122880.1
-
1
766
HQ122881.1
-
1
766
HQ122882.1
-
1
766
HQ122883.1
-
1
766
HQ122885.1
-
1
766
HQ122886.1
-
1
766
HQ122891.1
-
1
766
HQ122892.1
-
1
766
HQ122893.1
-
1
766
HQ122894.1
-
1
766
HQ122895.1
-
1
766
HQ122896.1
-
1
766
HQ122897.1
-
1
766
HQ122898.1
-
1
766
HQ122899.1
-
1
766
HQ122900.1
-
1
766
HQ122901.1
-
1
767
HQ122902.1
-
1
766
HQ122903.1
-
1
766
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0