Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004581.5 | NP_004572.3 | 10 | 1552 | ||
NM_182836.2 | NP_878256.1 | 11 | 1235 | |||
GenBank mRNA | AK291347.1 | - | 11 | 1119 | ||
AK292613.1 | - | 10 | 1284 | |||
AK303247.1 | - | 9 | 997 | |||
AK308715.1 | - | 11 | 1118 | |||
BC003093.1 | - | 11 | 1173 | 3537388 | ||
CR541797.1 | - | 9 | 1040 | |||
Y08200.1 | - | 10 | 1314 |