Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005059.3 | NP_005050.2 | 3 | 370 | ||
NM_134441.2 | NP_604390.1 | 2 | 269 | |||
XM_017015011.1 | XP_016870500.1 | 3 | 638 | |||
GenBank mRNA | A06925.1 | - | 2 | 238 | ||
A17315.1 | - | 2 | 238 | |||
AA935560.1 | - | 3 | 191 | |||
BC126415.1 | - | 2 | 269 | 8992131 | ||
BC126419.1 | - | 2 | 269 | 8992135 | ||
X00948.1 | - | 2 | 238 |