Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014.4 | NP_001005.1 | 4 | 435 | ||
NM_001203245.2 | NP_001190174.1 | 4 | 587 | |||
NM_001204091.1 | NP_001191020.1 | 4 | 439 | |||
GenBank mRNA | AB209165.1 | - | 1 | 390 | ||
AK311797.1 | - | 4 | 382 | |||
BC001032.2 | - | 4 | 360 | 3139393 | ||
BC001955.1 | - | 4 | 414 | 2905318 | ||
BC005012.1 | - | 4 | 414 | 3637658 | ||
BC070235.1 | - | 4 | 358 | 6453788 | ||
BC071946.1 | - | 4 | 370 | 6305105 | ||
BC073799.1 | - | 4 | 447 | 5517455 | ||
BE397113.1 | - | 4 | 385 | |||
BQ072109.1 | - | 1 | 555 | |||
BU956627.1 | - | 4 | 363 | |||
BY996439.1 | - | 4 | 594 | |||
CD385233.1 | - | 4 | 437 | |||
U14972.1 | - | 1 | 357 | |||
Z70761.1 | - | 1 | 23 |
Gene Symbol: RPS10-NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202470.2 | NP_001189399.1 | 4 | 435 | ||
GenBank mRNA | BU521288.1 | - | 4 | 358 |
Gene Symbol: RPS10P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025353.1 | - | 1 | 2969 |