Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SCNN1D SIRNA Assays ›
Gene Symbol:
SCNN1D
Gene Name:
sodium channel epithelial 1 delta subunit
Gene Aliases:
ENaCd, ENaCdelta, SCNED, dNaCh
Chromosome Location:
Chr.1: 1280415 - 1292029 on Build GRCh38
UniGene ID:
Hs.512681
Species:
Human
siRNA ID
HSS143823
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SCNN1D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130413.3
NP_001123885.2
13
1939
NR_037668.2
12
1845
XM_011541899.2
XP_011540201.1
13
2609
XM_011541901.2
XP_011540203.1
14
2131
XM_011541902.2
XP_011540204.1
14
2107
XM_011541905.2
XP_011540207.1
13
2035
XM_011541906.2
XP_011540208.1
10
2369
XM_011541908.2
XP_011540210.1
13
2306
XM_011541920.2
XP_011540222.1
12
2129
XM_011541925.2
XP_011540227.1
11
1688
XM_011541929.2
XP_011540231.1
9
2497
XM_011541932.2
XP_011540234.1
10
1594
XM_011541933.2
XP_011540235.1
10
1560
XM_017002037.1
XP_016857526.1
12
2606
XM_017002038.1
XP_016857527.1
11
2459
XM_017002039.1
XP_016857528.1
13
1984
XM_017002040.1
XP_016857529.1
13
1846
XM_017002041.1
XP_016857530.1
14
1834
XM_017002042.1
XP_016857531.1
13
1825
XM_017002043.1
XP_016857532.1
13
1801
XM_017002044.1
XP_016857533.1
12
1750
GenBank mRNA
AK093239.1
-
10
1569
AK093372.1
-
7
694
AK127357.1
-
10
2365
AK131558.1
-
14
2265
AK315288.1
-
9
1416
BC036752.1
-
9
1388
BC040882.1
-
12
1809
BC125074.1
-
9
1338
40117641
BC125075.1
-
8
1440
40117643
DQ898175.1
-
9
1313
DQ898176.1
-
10
1419
EU489064.1
-
8
1221
U38254.1
-
8
1221
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
sodium ion transport
ion transmembrane transport
sodium ion transmembrane transport
response to stimulus
sensory perception of taste
protein binding
ligand-gated sodium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0