Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005623.2 | NP_005614.2 | 2 | 562 | ||
GenBank mRNA | BC126242.1 | - | 2 | 369 | 8991958 | |
Y10802.1 | - | 2 | 163 | |||
Y16645.1 | - | 2 | 579 |