Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002989.3 | NP_002980.1 | 3 | 436 | ||
XM_011518004.2 | XP_011516306.1 | 3 | 423 | |||
GenBank mRNA | AB002409.1 | - | 3 | 393 | ||
AF001979.1 | - | 3 | 335 | |||
AK311901.1 | - | 3 | 405 | |||
AY358887.1 | - | 3 | 340 | |||
BC027918.1 | - | 3 | 342 | 5226781 | ||
BI833188.1 | - | 3 | 418 | |||
BP331359.1 | - | 3 | 436 | |||
CR450326.1 | - | 3 | 335 | |||
U88320.1 | - | 3 | 349 |