Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000542.3 | NP_000533.3 | 9 | 1084 | ||
NM_198843.2 | NP_942140.2 | 9 | 1084 | |||
XM_005264487.2 | XP_005264544.1 | 8 | 999 | |||
XM_005264488.3 | XP_005264545.2 | 8 | 952 | |||
XM_005264490.4 | XP_005264547.2 | 8 | 964 | |||
XM_017004702.1 | XP_016860191.1 | 8 | 999 | |||
XM_017004703.1 | XP_016860192.1 | 8 | 952 | |||
GenBank mRNA | AK303817.1 | - | 7 | 872 | ||
AK314947.1 | - | 8 | 964 | |||
BC032785.1 | - | 8 | 951 | 5179408 | ||
DQ317589.1 | - | 8 | 925 | |||
J02761.1 | - | 8 | 961 | |||
M19097.1 | - | 4 | 534 |