Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMTN SIRNA Assays ›
Gene Symbol:
SMTN
Gene Name:
smoothelin
Gene Aliases:
-
Chromosome Location:
Chr.22: 31081296 - 31104624 on Build GRCh38
UniGene ID:
Hs.149098
Species:
Human
siRNA ID
HSS143957
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001207017.1
NP_001193946.1
11
1718
NM_001207018.1
NP_001193947.1
10
1740
NM_006932.4
NP_008863.3
11
1736
NM_134269.2
NP_599031.1
11
1736
NM_134270.2
NP_599032.2
11
1736
XM_006724298.1
XP_006724361.1
11
1714
XM_006724302.1
XP_006724365.1
11
1773
XM_011530333.2
XP_011528635.1
12
1819
XM_011530336.2
XP_011528638.1
12
1820
XM_011530337.2
XP_011528639.1
12
1820
XM_011530338.2
XP_011528640.1
12
1819
XM_011530339.2
XP_011528641.1
12
1820
XM_011530340.2
XP_011528642.1
12
1820
XM_011530341.1
XP_011528643.1
4
651
XM_017028913.1
XP_016884402.1
12
1820
XM_017028914.1
XP_016884403.1
12
1819
XM_017028915.1
XP_016884404.1
12
1819
XM_017028916.1
XP_016884405.1
12
1820
XM_017028917.1
XP_016884406.1
12
1820
XM_017028918.1
XP_016884407.1
12
1820
XM_017028919.1
XP_016884408.1
12
1819
GenBank mRNA
AB209643.1
-
8
1309
AF064238.3
-
11
1713
AJ010306.2
-
11
1713
AK096953.1
-
11
1771
AK128627.1
-
10
3281
AK128719.1
-
11
1582
AK225648.1
-
2
177
AK295418.1
-
10
1740
AK304084.1
-
11
1718
AK307627.1
-
2
185
AY061971.1
-
11
1714
AY061972.1
-
11
1714
BC034237.2
-
11
1704
3912887
Y13492.2
-
11
1714
Z49989.2
-
2
170
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
smooth muscle contraction
muscle organ development
actin binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0