Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC8A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015063.2 | NP_055878.1 | 3 | 1034 | ||
XM_005259172.1 | XP_005259229.1 | 3 | 843 | |||
GenBank mRNA | AB029010.1 | - | 3 | 849 | ||
AK293860.1 | - | 2 | 167 | |||
AK302552.1 | - | 3 | 1034 | |||
AL512764.1 | - | 2 | 144 |