Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204194.1 | NP_001191123.1 | 3, 4 | 408 | ||
NM_001204196.1 | NP_001191125.1 | 3, 4 | 404 | |||
NM_003135.2 | NP_003126.1 | 4, 5 | 480 | |||
GenBank mRNA | AK311803.1 | - | 4, 5 | 388 | ||
BC000186.1 | - | 4, 5 | 372 | |||
BC010947.1 | - | 4, 5 | 364 | 4285753 | ||
BU661702.1 | - | 3, 4 | 282 | |||
DN915735.1 | - | 3, 4 | 273 | |||
X12791.1 | - | 4, 5 | 372 |
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BE047584.1 | - | 7, 8 | 505 |